Order Entry
Poland
Orders LinkContactUsLinkComponent
Seals for threaded connection caps (Septa)

Seals for threaded connection caps (Septa)

GL45 closure size, 42 mm, 3 mm
Catalog # 201-1965
Supplier:   BIRZLE PETER
Seals for threaded connection caps (Septa)
Catalog # 201-1965
Supplier:  BIRZLE PETER
Seals for threaded connection caps (Septa)
Seals for threaded connection caps (Septa)

GL45 closure size, 42 mm, 3 mm

Price
8,80 zł
Log in for availability
1 * 1 SZT
Product Details & Documents

Silicone, transparent.

  • Ideal for use as septa (for puncturing)
  • Very good chemical resistance
  • Very good thermal resistance

For threaded connection caps with GL thread.

Specifications
  • Ø:
    42 mm
  • For:
    GL45 closure size
  • Seq:
    04
  • Thickness:
    3 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.