Kolby filtracyjne do systemów filtracyjnych, Nalgene™ Rapid-Flow™
Product Details & Documents
Save time and minimize contamination with gamma-sterilized Thermo Scientific™ Nalgene™ Rapid-Flow™ Filter Storage Bottles made from virgin PS. Use with Nalgene Bottle-Top filters or as replacement/extra receptacles for filter units. Excellent as single-use storage containers.
Headspace allows for extra serum or other additives to be added to the filtrate
Leakproof screw caps to eliminate pH shift in bottle for longer life of stored media
Sterilised by gamma radiation and individually bagged for a 5-year sterile shelf life
Non-pyrogenic and non-cytotoxic
Cap is made from virgin HDPE.
Specifications
- Pojemność:250 ml
- Seq:02
- Cat. No.:734-5086
- Environmentally Preferable:Niski wpływ na produkcję
- Certyfikacja środowiskowa:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)