2-(Bromomethyl)-1,3-dioxolane 97%
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
2-(Bromomethyl)-1,3-dioxolane 97%
Catalog # ACRO209841000
Supplier: Thermo Fisher Scientific
CAS Number: 4360-63-8
Jump to:
Product Details & Documents
Synonyms: 2-Bromomethyl-1,3-dioxolane
- CAS Number:4360-63-8
- Formula:C₄H₇BrO₂
- MDL Number:MFCD00003214
- Molecular Weight:167 g/mol
- Boiling Pt:80…82 °C (27 mmHg)
- Flash Pt:62 °C
- Storage Temperature:Ambient
- EINECS:224-443-6
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4810 to 1.4840 (20°C, 589 nm)
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:100 mL



