Copper(I) oxide 97%
Glass bottle for solids, 500 g
Dostawca: Thermo Fisher Scientific
Copper(I) oxide 97%
numer katalogowy ACRO197425000
Dostawca: Thermo Fisher Scientific
Numer CAS: 1317-39-1
Przejdź do:
Product Details & Documents
Synonimy: Cuprous oxide, Dicopper oxide
- Numer CAS:1317-39-1
- Wzór:Cu₂O
- Numer MDL:MFCD00010974
- Molecular Weight:143.09 g/mol
- Indeks Mercka:13,02694
- Temperatura wrzenia:1800 °C (1013 hPa)
- Temperatura topnienia:1232 °C
- Gęstość:5,7 to 6.2 g/cm³ (20 °C)
- Temperatura przechowywania:Ambient
- EINECS:215-270-7
- UN:3077
- ADR:9,III
Documents
ACRO19742
SDS
Dane techniczne
Wyniki testu specyfikacji
- Appearance:Red-brown Powder
- Titration Complexometric:≥96.0 %
- Iron (Fe):≤500 ppm
- Nickel (Ni):≤100 ppm
- Silver (Ag):≤50 ppm
Dane techniczne
- Pack type:Glass bottle for solids
- Pk:500 g
Dane techniczne
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



