Lactobionic acid 97%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoic acid, 4-O-β-D-Galactopyranosyl-D-gluconic acid
- CAS Number:96-82-2
- Formula:C₁₂H₂₂O₁₂
- MDL Number:MFCD00078147
- Molecular Weight:358.30 g/mol
- Melting Pt:113…118 °C
- Storage Temperature:Ambient
Documents
ACRO16711
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤5 %
- Heavy metals:≤10 ppm
- Sulfated ash:≤0.1 %
- Specific optical rotation:+22.8° to +28.8° (20°C, 589 nm) (c=10, water) (after 4 h)
- Titration with NaOH:≥96.0 %
- Chloride (Cl):≤400 ppm
- Sulfate (SO4):≤400 ppm
- Arsenic (As):≤2 ppm
- Calcium (Ca):≤500 ppm
- Iron (Fe):≤80 ppm
Specifications
- Pack type:Plastic bottle
- Pk:100 g



