Lactobionic acid 97%
Plastic bottle, 25 g
Dostawca: Thermo Fisher Scientific
Lactobionic acid 97%
numer katalogowy ACRO167110250
Dostawca: Thermo Fisher Scientific
Numer CAS: 96-82-2
Jump to:
Product Details & Documents
Synonyms: (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoic acid, 4-O-β-D-Galactopyranosyl-D-gluconic acid
- CAS Number:96-82-2
- Formula:C₁₂H₂₂O₁₂
- MDL Number:MFCD00078147
- Molecular Weight:358.30 g/mol
- Melting Pt:113…118 °C
- Storage Temperature:Ambient
Documents
ACRO16711
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤5 %
- Heavy metals:≤10 ppm
- Sulfated ash:≤0.1 %
- Specific optical rotation:+22.8° to +28.8° (20°C, 589 nm) (c=10, water) (after 4 h)
- Titration with NaOH:≥96.0 %
- Chloride (Cl):≤400 ppm
- Sulfate (SO4):≤400 ppm
- Arsenic (As):≤2 ppm
- Calcium (Ca):≤500 ppm
- Iron (Fe):≤80 ppm
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Cat. No.:ACRO167110250
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



