1,5-Dibromopentane 97%
Glass bottle, 250 mL
Dostawca: Thermo Fisher Scientific
1,5-Dibromopentane 97%
numer katalogowy ACRO155992500
Dostawca: Thermo Fisher Scientific
Numer CAS: 111-24-0
Przejdź do:
Product Details & Documents
- Numer CAS:111-24-0
- Wzór:BrH₂C(CH₂)₃CH₂Br
- Numer MDL:MFCD00000268
- Molecular Weight:229.94 g/mol
- Temperatura wrzenia:224 °C (1013 hPa)
- Temperatura topnienia:–34 °C
- Gęstość:1.68 g/cm³
- Punkt zapalny:79 °C
- Temperatura przechowywania:Ambient
- EINECS:203-849-7
Documents
ACRO15599
SDS
Dane techniczne
Wyniki testu specyfikacji
- Appearance:Clear colorless to yellow-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5112 to 1.5132 (20°C, 589 nm)
- GC:≥96.0 %
Dane techniczne
- Pack type:Glass bottle
- Pk:250 mL
Dane techniczne
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



