1,2-Dibromo-1,2-diphenylethane 96%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
1,2-Dibromo-1,2-diphenylethane 96%
Catalog # ACRO147950050
Supplier: Thermo Fisher Scientific
CAS Number: 5789-30-0
Jump to:
Product Details & Documents
- CAS Number:5789-30-0
- Formula:C₆H₅(CHBr)₂C₆H₅
- MDL Number:MFCD00000137
- Molecular Weight:340.06 g/mol
- Melting Pt:235…241 °C (dec.)
- Storage Temperature:Ambient
- EINECS:227-324-7
- UN:3261
- ADR:8,III
Documents
ACRO14795
SDS
Specifications
Specification Test Results
- Appearance:White to yellow to light orange Powder
- Infrared spectrum:Conforms
- Melting point:234 to 241 °C
- HPLC:≥95 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



