4'-Nitroacetophenone 97%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
4'-Nitroacetophenone 97%
Catalog # ACRO128341000
Supplier: Thermo Fisher Scientific
CAS Number: 100-19-6
Jump to:
Product Details & Documents
Synonyms: p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
- CAS Number:100-19-6
- Formula:C₈H₇NO₃
- MDL Number:MFCD00007355
- Molecular Weight:165.15 g/mol
- Boiling Pt:202 °C (760 mmHg)
- Melting Pt:76…80 °C
- Flash Pt:149 °C
- Storage Temperature:Ambient
- EINECS:202-827-4
Documents
ACRO12834
SDS
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:76 to 80 °C
- Water:≤0.6 %
- GC:≥96.0 %
- Free acid:≤0.2 % (as 4-Nitrobenzoic acid)
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



