Order Entry
Poland
Orders LinkContactUsLinkComponent
 

4'-Nitroacetophenone 97%

Glass bottle for solids, 5 g
  ACRO128340050
 
4'-Nitroacetophenone 97%
  ACRO128340050
 :  100-19-6
4'-Nitroacetophenone 97%
4'-Nitroacetophenone 97%

Glass bottle for solids, 5 g

 :  100-19-6
 
181 zł
   
1 * 5 g
 
 :  p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
  •  :100-19-6
  •  :C₈H₇NO₃
  •  :MFCD00007355
  •  :165.15 g/mol
  •  :202 °C (760 mmHg)
  •  :76…80 °C
  •  :149 °C
  •  :Ambient
  •  :202-827-4

 

 

 

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    76 to 80 °C
  • Water:
    ≤0.6 %
  • GC:
    ≥96.0 %
  • Free acid:
    ≤0.2 % (as 4-Nitrobenzoic acid)

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g