Methyl-4-hydroxybenzoate 99%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Methyl-4-hydroxybenzoate 99%
Catalog # ACRO126961000
Supplier: Thermo Fisher Scientific
CAS Number: 99-76-3
Jump to:
Product Details & Documents
Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
- CAS Number:99-76-3
- Formula:HOC₆H₄CO₂CH₃
- MDL Number:MFCD00002352
- Molecular Weight:152.15 g/mol
- Merck Index:13,06129
- Boiling Pt:298.6 °C (760 mmHg)
- Melting Pt:125…128 °C
- Density:1,1208 g/cm³ (137 °C)
- Storage Temperature:Ambient
- EINECS:202-785-7
Documents
Specifications
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:125 to 128 °C
- HPLC:≥98.5 %
- Residue after ignition:≤0.1 %
Specifications
- Pack type:Plastic bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



