Order Entry
Poland
Orders LinkContactUsLinkComponent
 

1-Hexadecanol 96%

Plastic drum, 2,5 kg
Catalog # ACRO120480025
Irritant
1-Hexadecanol 96%
Catalog # ACRO120480025
CAS Number:  36653-82-4
1-Hexadecanol 96%
1-Hexadecanol 96%

Plastic drum, 2,5 kg

CAS Number:  36653-82-4
Price
643 zł
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Cetyl alcohol, Palmityl alcohol
  • CAS Number:36653-82-4
  • Formula:H₃C(CH₂)₁₄CH₂OH
  • MDL Number:MFCD00004760
  • Molecular Weight:242.45 g/mol
  • Merck Index:13,02037
  • Boiling Pt:344 °C (760 mmHg)
  • Melting Pt:47…50 °C
  • Density:0,81 g/cm³ (20 °C)
  • Flash Pt:>157 °C
  • Storage Temperature:Ambient
  • EINECS:253-149-0
Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder or flakes or pellets
  • Infrared spectrum:
    Conforms
  • GC:
    ≥95 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    2,5 kg

Specifications