p-Benzoquinone 99%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone
- CAS Number:106-51-4
- Formula:C₆H₄O₂
- MDL Number:MFCD00001591
- Molecular Weight:108.1 g/mol
- Merck Index:12,08259
- Boiling Pt:~ 174 °C (1013 hPa)
- Melting Pt:112…116 °C
- Density:1.31 g/cm³
- Flash Pt:77 °C
- Storage Temperature:Ambient
- EINECS:203-405-2
- UN:2587
- ADR:6.1,II
Documents
Specifications
Specification Test Results
- Appearance:Yellow to green Powder
- Infrared spectrum:Conforms
- Melting point:112 to 116 °C
- Titration Iodometric:≥98.5 %
- Water:≤0.5 %
- Specific optical rotation:-218° to -224° (20 °C, 589 nm) (c=2, 1M HCl) (On dried substance)
- Titration:98.5 to 101.0 % (On dried substance)
- Sulfates:≤300 ppm
- Chlorides:≤200 ppm
- Ammonium (NH4):≤200 ppm
- Iron (Fe):≤10 ppm
- Ninhydrin-positive substances:Passes test
Specifications
- Pack type:Plastic bottle
- Pk:500 g



