Benzaldehyde 98+%
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:100-52-7
- Formula:C₆H₅CHO
- MDL Number:MFCD00003299
- Molecular Weight:106.12 g/mol
- Merck Index:13,01057
- Boiling Pt:179 °C (1013 hPa)
- Melting Pt:–26 °C
- Density:1.043 g/cm³
- Flash Pt:64 °C
- Storage Temperature:Ambient
- EINECS:202-860-4
- UN:1990
- ADR:9,III
Documents
ACRO10522
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5440 to 1.5460 (20°C, 589 nm)
- GC:≥98.0 %
- Specific gravity:(25°C) 1.041 to 1.046
- Acid value:≤5 mg KOH/g
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



