1,2-Dibromo-2-methylpropane 98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
1,2-Dibromo-2-methylpropane 98%
Catalog # ACRO406490250
Supplier: Thermo Fisher Scientific
CAS Number: 594-34-3
Jump to:
Product Details & Documents
- CAS Number:594-34-3
- Formula:C₄H₈Br₂
- MDL Number:MFCD00000126
- Molecular Weight:215.92 g/mol
- Boiling Pt:150 °C (760 mmHg)
- Melting Pt:-70,1 °C
- Density:1.79 g/cm³
- Flash Pt:>110 °C
- :Density: 1.7675 g/cm³ (0 °C)
- Storage Temperature:Ambient
- EINECS:209-835-7
Documents
ACRO40649
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5083 to 1.5103 (20°C, 589 nm)
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



