Order Entry
Poland
Orders LinkContactUsLinkComponent
Mortar spatulas

Mortar spatulas

200 mm, 40 mm, 20 mm
Catalog # BOCH3261
Supplier:   BOCHEM
Mortar spatulas
Catalog # BOCH3261
Supplier:  BOCHEM
Mortar spatulas
Mortar spatulas

200 mm, 40 mm, 20 mm

Price
78,20 zł
Log in for availability
1 * 1 SZT
Product Details & Documents

Double-ended mortar spatulas made of stainless steel.

Specifications
  • Length:
    200 mm
  • Blade width:
    20 mm
  • Blade length:
    40 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.