1,8-Octanedithiol 99%
Glass bottle for solids, 5 g
: 1,8-Dimercaptooctane
- :1191-62-4
- :C₈H₁₈S₂
- :MFCD00003574
- :178.36 g/mol
- :269…270 °C (760 mmHg)
- :0.96 g/cm³
- :>110 °C
- :Ambient
- :214-738-8
- :3334
- :9,III
- :Density: 0.9814 g/cm³ (0 °C)
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5030 to 1.5050 (20°C, 589 nm)
- GC:≥98.5 %
- Pack type:Glass bottle for solids
- Pk:5 g



