Order Entry
Poland
Orders LinkContactUsLinkComponent
 

(2-Hydroxylpropyl)-β-cyclodextrine 97%

Glass bottle, 25 g
  ACRO297560250
(2-Hydroxylpropyl)-β-cyclodextrine 97%
  ACRO297560250
 :  128446-35-5
(2-Hydroxylpropyl)-β-cyclodextrine 97%
(2-Hydroxylpropyl)-β-cyclodextrine 97%

Glass bottle, 25 g

 :  128446-35-5
 
580 zł
   
1 * 25 g
 
 :  Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
  •  :128446-35-5
  •  :C₆₃H₁₁₂O₄₂
  •  :MFCD16621721
  •  :278 °C (dec.)
  •  :Ambient
 

 

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤7.5 % (K.F.)
  • HPLC:
    ≥96.0 % (On dry substance)
  • NMR:
    2 to 6 (degree of substitution)

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g