Order Entry
Poland
Orders LinkContactUsLinkComponent
Pipette controller, pipetus®-standard

Pipette controller, pipetus®-standard

pipetus® standard, with pipette holder, safety valve and filter, 120 V, US-plug
Catalog # HIRS9905000
Pipette controller, pipetus®-standard
Catalog # HIRS9905000
Supplier:  
Pipette controller, pipetus®-standard
Pipette controller, pipetus®-standard

pipetus® standard, with pipette holder, safety valve and filter, 120 V, US-plug

Product Details & Documents

Pipette controller for pipettes from 0,1 to 100 ml.

  • Optimum safety and excellent convenience of use
  • Pipettor is lighter and easy to use because the pump is separated from the handle
  • Autoclavable, detachable handle for the pipette holder is equipped with two buttons for filling and emptying

This electric pipettor is equipped with a 0,2 µm integrated water-repellent filter (for better durability) and a safety valve to protect the equipment from the introduction of hazardous (contaminated or radioactive) liquids.

Ordering information: The complete standard pipetus® is supplied with an electric pump, two filters and a safety valve.

Specifications
  • Description:
    pipetus® standard, with pipette holder, safety valve and filter, 120 V, US-plug
  • Seq:
    05
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • External pump with filter, 230 V, EU
    External pump with filter, 230 V, EU
    HIRS9904400
    Log in for availability
    1 * 1 SZT - 876 zł

Product Information

  • Catalog #
    Availability
    Price
  • Silicone adapter
    Silicone adapter
    612-2615
    Log in for availability
    1 * 1 SZT - 41,70 zł

Product Information

  • Catalog #
    Availability
    Price
  • Round filter set with one-sided connection (50 filters)
    Round filter set with one-sided connection (50 filters)
    HIRS9903705
    Log in for availability
    1 * 50 SZT - 1 480 zł

Product Information

  • Catalog #
    Availability
    Price
  • Safety valve
    Safety valve
    612-2616
    Log in for availability
    1 * 1 SZT - 155 zł

Product Information

  • Catalog #
    Availability
    Price
  • Microfibre-filter set for pump (5 filters)
    Microfibre-filter set for pump (5 filters)
    612-2618
    Log in for availability
    1 * 5 SZT - 3 400 zł

Product Information

  • Catalog #
    Availability
    Price
  • Spare O-rings for pipetus® akku and pipetus®
    Spare O-rings for pipetus® akku and pipetus®
    HIRS9903205
    Log in for availability
    1 * 5 SZT - 201 zł

Product Information

  • Catalog #
    Availability
    Price
  • Pipette holder, complete, turquoise, for pipetus®/pipetus® accu/pipetus® standard/pipetus® classic
    Pipette holder, complete, turquoise, for pipetus®/pipetus® accu/pipetus® standard/pipetus® classic
    HIRS9903300
    Log in for availability
    1 * 1 SZT - 295 zł

Product Information

  • Catalog #
    Availability
    Price
  • Round filter set with one-sided connection (5 filters)
    Round filter set with one-sided connection (5 filters)
    612-1982
    Log in for availability
    1 * 5 SZT - 170 zł

Product Information

  • Catalog #
    Availability
    Price
  • Filter set, sterile, individually packed, for pipetus®
    Filter set, sterile, individually packed, for pipetus®
    HIRS9903714
    Log in for availability
    1 * 5 SZT - 1 070 zł

Product Information

  • Catalog #
    Availability
    Price
  • Round filter set with one-sided connection (100 filters)
    Round filter set with one-sided connection (100 filters)
    HIRS9903706
    Log in for availability
    1 * 100 SZT - 2 560 zł