Order Entry
Poland
Orders LinkContactUsLinkComponent
 

LC/MS calibrant mixes

Electrospray ionisation (ESI) tuning mix, 100 ml, 100 mL
  HEWLG2421-60001
 :   Agilent
LC/MS calibrant mixes
  HEWLG2421-60001
 :  Agilent
 :  
LC/MS calibrant mixes
LC/MS calibrant mixes

Electrospray ionisation (ESI) tuning mix, 100 ml, 100 mL

 :  
 
1 520 zł
   
1 * 100 mL
 

 

 

 

  • Pk:
    100 mL
  • Description:
    Electrospray ionisation (ESI) tuning mix, 100 ml