Microvolume inserts for screw top vials
Flat bottom, 0,20 ml, Glass, No
Catalog # HEWL5183-2090
Supplier: Agilent
Jump to:
Product Details & Documents
Specifications
- Cap included:No
- Capacity:0,20 ml
- Description:Flat bottom
- Material:Glass
- Seq:03