Top tilt measures
10 ml
Catalog # HECH41877006
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Automatic tilt measures, made of DURAN® borosilicate glass 3.3, with standard ground cone NS 29/32. Supplied without reservoir bottle.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
- Capacity:10 ml
- Seq:02