Order Entry
Poland
Orders LinkContactUsLinkComponent
pH Buffer solutions in bottles

pH Buffer solutions in bottles

pH Buffer solution with certificates of analysis, 4,01, ±0,01, Colourless, 25 °C, 500 mL
Catalog # HANNHI7004L/C
Supplier:   Hanna
pH Buffer solutions in bottles
Catalog # HANNHI7004L/C
Supplier:  Hanna
CAS Number:  
pH Buffer solutions in bottles

pH Buffer solution with certificates of analysis, 4,01, ±0,01, Colourless, 25 °C, 500 mL

CAS Number:  
Price
247 zł
Log in for availability
1 * 500 mL
Product Details & Documents

Plastic bottle with a tamper-proof seal.

  • Color coded label

Traceable to NIST standard reference materials.

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Colourless
  • Description:
    pH Buffer solution with certificates of analysis
  • pH accuracy:
    ±0,01
  • pH value:
    4,01

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.