Biology prepared microscope slides
Pine (Pinus) female cone with ovules longitudinal section
LIED747D
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
- Microtome sections individually crafted under rigorous scientific control
- Each slide carefully selected on the basis of instructional value
- Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
: Supplied on high quality ground edge slides (26×76 mm).
- Description:Pine (Pinus) female cone with ovules longitudinal section
- Seq:22