Order Entry
Poland
Orders LinkContactUsLinkComponent
Biology prepared microscope slides
Biology prepared microscope slides
Foliose lichen (Physcia) thallus with symbiotic algae transverse section
Catalog # LIED735D
Supplier:   LIEDER JOHANNES
Biology prepared microscope slides
Catalog # LIED735D
Supplier:  LIEDER JOHANNES
Biology prepared microscope slides
Biology prepared microscope slides
Foliose lichen (Physcia) thallus with symbiotic algae transverse section
Price
37,90 zł
Log in for availability
1 * 1 SZT
Product Details & Documents
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
 : Supplied on high quality ground edge slides (26×76 mm).
Specifications
  • Description:
    Foliose lichen (Physcia) thallus with symbiotic algae transverse section
  • Seq:
    10