Order Entry
Poland
Orders LinkContactUsLinkComponent
Biology prepared microscope slides | Avantor
Biology prepared microscope slides
Retina of cat with rods and cones
numer katalogowy LIED723D
Dostawca:   LIEDER JOHANNES
Biology prepared microscope slides
numer katalogowy LIED723D
Dostawca:  LIEDER JOHANNES
Biology prepared microscope slides
Biology prepared microscope slides
Retina of cat with rods and cones
Przejdź do:
Price
37,90 zł
Zaloguj się dla dostępności
1 * 1 SZT
Product Details & Documents
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
  • Microtome sections individually crafted under rigorous scientific control
  • Each slide carefully selected on the basis of instructional value
  • Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
Informacja o dostawie: Supplied on high quality ground edge slides (26×76 mm).
Dane techniczne
  • Description:
    Retina of cat with rods and cones
  • Seq:
    28