Biology prepared microscope slides
Ovary of cat with primary, secondary and Graafian follicles
LIED718D
The slides are arranged and compiled, so that each enlarges the subject line of the proceeding one.
- Microtome sections individually crafted under rigorous scientific control
- Each slide carefully selected on the basis of instructional value
- Multicolour staining for distinct variation
The set contains slides of typical micro-organisms, cell division and embryonic developments, as well as of tissues and organs of plants, animals and man.
: Supplied on high quality ground edge slides (26×76 mm).
- Description:Ovary of cat with primary, secondary and Graafian follicles
- Seq:21