Order Entry
Poland
Orders LinkContactUsLinkComponent
WHEATON® Vials with Inserts

WHEATON® Vials with Inserts

Autosampler vial, with 0,3 ml PP insert, 0,30 ml, 12×32 mm, No, Snap/crimp
Catalog # WHEA225180
Supplier:   DWK Life Sciences
WHEATON® Vials with Inserts
Catalog # WHEA225180
Supplier:  DWK Life Sciences
WHEATON® Vials with Inserts
WHEATON® Vials with Inserts

Autosampler vial, with 0,3 ml PP insert, 0,30 ml, 12×32 mm, No, Snap/crimp

Price
1 520 zł
Log in for availability
1 * 1.000 SZT
Product Details & Documents

These vials are made of durable PP and can be used in standard gas and liquid chromatography autosamplers.

  • Lightweight and economical
  • Available with snap/crimp or screw top finish
  • Conical interior ensures maximum retrieval of contents without the hassle of using removable inserts

Packaging: 100 vials per shelf pack or 1000 vials per case.

Specifications
  • Cap included:
    No
  • Ø×H:
    12×32 mm
  • Capacity:
    0,30 ml
  • Description:
    Autosampler vial, with 0,3 ml PP insert
  • Seq:
    01
  • Cap type:
    Snap/crimp