Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Copper(I) oxide 97%

Bottle
Catalog # APOSIN10415-500G
New Product
Supplier:   Apollo Scientific
Copper(I) oxide 97%
Catalog # APOSIN10415-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  1317-39-1
Copper(I) oxide 97%

Bottle

CAS Number:  1317-39-1
Price
319 zł
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Inorganic salt or catalyst

Synonyms:  Cuprous oxide, Dicopper oxide
  • CAS Number:1317-39-1
  • Formula:Cu₂O
  • MDL Number:MFCD00010974
  • Molecular Weight:143,09 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications