Order Entry
Poland
Orders LinkContactUsLinkComponent
Copper(I) oxide 97%

Copper(I) oxide 97%

Bottle
Catalog # APOSIN10415-100G
New Product
Supplier:   Apollo Scientific
Copper(I) oxide 97%
Catalog # APOSIN10415-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  1317-39-1
Copper(I) oxide 97%

Bottle

CAS Number:  1317-39-1
Price
114 zł
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Inorganic salt or catalyst

Synonyms:  Cuprous oxide, Dicopper oxide
  • CAS Number:1317-39-1
  • Formula:Cu₂O
  • MDL Number:MFCD00010974
  • Molecular Weight:143,09 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.