Order Entry
Poland
Orders LinkContactUsLinkComponent
Copper(I) oxide 97%
Copper(I) oxide 97%
Bottle
  APOSIN10415-25G
New Product
Copper(I) oxide 97%
  APOSIN10415-25G
New Product
 :  1317-39-1
Copper(I) oxide 97%
Bottle
 :  1317-39-1
107 zł
1 * 25 g
Usage: Inorganic salt or catalyst
 :  Cuprous oxide, Dicopper oxide
  •  :1317-39-1
  •  :Cu₂O
  •  :MFCD00010974
  •  :143,09 g/mol
  • Pack type:
    Bottle