Order Entry
Poland
Orders LinkContactUsLinkComponent
2-Octanol ≥98%

2-Octanol ≥98%

Bottle
Catalog # APOSBUP13780-50MG
New Product
Supplier:   Apollo Scientific
2-Octanol ≥98%
Catalog # APOSBUP13780-50MG
Supplier:  Apollo Scientific
New Product
CAS Number:  123-96-6
2-Octanol ≥98%

Bottle

CAS Number:  123-96-6
Price
418 zł
Log in for availability
1 * 50 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
  • CAS Number:123-96-6
  • Formula:C₈H₁₈O
  • Molecular Weight:130,23 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications