2,2-Dichloroacetamide
Bottle
Catalog # APOSOR14320-500G
New Product
Supplier: Apollo Scientific
2,2-Dichloroacetamide
Catalog # APOSOR14320-500G
Supplier: Apollo Scientific
New Product
CAS Number: 683-72-7
Jump to:
Product Details & Documents
- CAS Number:683-72-7
- Formula:C₂H₃Cl₂NO
- MDL Number:MFCD00008015
- Molecular Weight:127.96 g/mol
- Boiling Pt:233…234 °C
- Melting Pt:96…101 °C
- Storage Temperature:Ambient
- EINECS:211-674-2
Specifications
Specifications
- Pack type:Bottle



