ML-9
Bottle
Catalog # APOSOR7351T-250MG
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Inhibits the catalytic activities of several protein linases competitively with respect to ATP. Also found to be a new type of vascular relaxant.
Synonyms: 1-(5-Chloronaphthalene-1-sulphonyl)-1H-hexahydro-1,4-diazepine hydrochloride, 1-((5-Chloronaphthalen-1-yl)sulphonyl)-1,4-diazepane hydrochloride
- CAS Number:105637-50-1
- Formula:C₁₅H₁₇N₂O₂SCl·HCl
- MDL Number:MFCD00065525
- Molecular Weight:361.3 g/mol
- Melting Pt:196…200 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



