ML-9
Bottle
Catalog # APOSOR7351T-100MG
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Inhibits the catalytic activities of several protein linases competitively with respect to ATP. Also found to be a new type of vascular relaxant.
Synonyms: 1-(5-Chloronaphthalene-1-sulphonyl)-1H-hexahydro-1,4-diazepine hydrochloride, 1-((5-Chloronaphthalen-1-yl)sulphonyl)-1,4-diazepane hydrochloride
- CAS Number:105637-50-1
- Formula:C₁₅H₁₇N₂O₂SCl·HCl
- MDL Number:MFCD00065525
- Molecular Weight:361.3 g/mol
- Melting Pt:196…200 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
Specifications
Rekomendacje
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



