Order Entry
Poland
Orders LinkContactUsLinkComponent
4,6-Dimethoxypyrimidine 97%

4,6-Dimethoxypyrimidine 97%

Bottle
Catalog # APOSOR912606-1G
New Product
Supplier:   Apollo Scientific
4,6-Dimethoxypyrimidine 97%
Catalog # APOSOR912606-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  5270-94-0
4,6-Dimethoxypyrimidine 97%

Bottle

CAS Number:  5270-94-0
Price
121 zł
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:5270-94-0
  • Formula:C₆H₈N₂O₂
  • MDL Number:MFCD00955844
  • Molecular Weight:140.14 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.