Order Entry
Poland
Orders LinkContactUsLinkComponent
Lactobionic acid ≥98%

Lactobionic acid ≥98%

Bottle
Catalog # APOSBUP16861-500MG
New Product
Supplier:   Apollo Scientific
Lactobionic acid ≥98%
Catalog # APOSBUP16861-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  96-82-2
Lactobionic acid ≥98%

Bottle

CAS Number:  96-82-2
Price
870 zł
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoic acid, 4-O-β-D-Galactopyranosyl-D-gluconic acid
  • CAS Number:96-82-2
  • Formula:C₁₂H₂₂O₁₂
  • Molecular Weight:358,3 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.