Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Lactobionic acid ≥98%

Bottle
  APOSBUP16861-250MG
New Product
Lactobionic acid ≥98%
  APOSBUP16861-250MG
New Product
 :  96-82-2
Lactobionic acid ≥98%

Bottle

 :  96-82-2
 
517 zł
   
1 * 250 mg
 

Usage: Biochemical/life science applications

 :  (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoic acid, 4-O-β-D-Galactopyranosyl-D-gluconic acid
  •  :96-82-2
  •  :C₁₂H₂₂O₁₂
  •  :358,3 g/mol
 

 

  • Pack type:
    Bottle