Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Lactobionic acid ≥98%

Bottle
Catalog # APOSBUP16861-100MG
New Product
Supplier:   Apollo Scientific
Lactobionic acid ≥98%
Catalog # APOSBUP16861-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  96-82-2
Lactobionic acid ≥98%

Bottle

CAS Number:  96-82-2
Price
262 zł
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoic acid, 4-O-β-D-Galactopyranosyl-D-gluconic acid
  • CAS Number:96-82-2
  • Formula:C₁₂H₂₂O₁₂
  • Molecular Weight:358,3 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications