Order Entry
Poland
Orders LinkContactUsLinkComponent
3,3-Dimethylcyclohexanone

3,3-Dimethylcyclohexanone

Bottle
Catalog # APOSOR315523-5G
Flammable
Corrosive
New Product
Supplier:   Apollo Scientific
3,3-Dimethylcyclohexanone
Catalog # APOSOR315523-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  2979-19-3
3,3-Dimethylcyclohexanone

Bottle

CAS Number:  2979-19-3
Price
121 zł
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:2979-19-3
  • Formula:C₈H₁₄O
  • MDL Number:MFCD00040178
  • Molecular Weight:126.2 g/mol
  • Boiling Pt:175 °C
  • Density:0.91 g/cm³ (25 °C)
  • Flash Pt:59 °C
  • Storage Temperature:Ambient
  • UN:1224
  • ADR:3,III
  •  :Density: 0.909 g/cm³ (25 °C)
Specifications

Specifications

  • Pack type:
    Bottle

Specifications