3,3-Dimethylindoline
Bottle
Catalog # APOSOR313014-25G
New Product
Supplier: Apollo Scientific
3,3-Dimethylindoline
Catalog # APOSOR313014-25G
Supplier: Apollo Scientific
New Product
CAS Number: 1914-02-9
Jump to:
Product Details & Documents
- CAS Number:1914-02-9
- Formula:C₁₀H₁₃N
- MDL Number:MFCD11847210
- Molecular Weight:147.22 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle



