Order Entry
Poland
Orders LinkContactUsLinkComponent
3,3-Dimethylindoline

3,3-Dimethylindoline

Bottle
Catalog # APOSOR313014-5G
Irritant
New Product
Supplier:   Apollo Scientific
3,3-Dimethylindoline
Catalog # APOSOR313014-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  1914-02-9
3,3-Dimethylindoline

Bottle

CAS Number:  1914-02-9
Price
2 400 zł
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:1914-02-9
  • Formula:C₁₀H₁₃N
  • MDL Number:MFCD11847210
  • Molecular Weight:147.22 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.