VWR® Socket Caps and Colour Inserts for Screw Microtubes
Customisable screw caps, non sterile
numer katalogowy 525-1671
New Product
Dostawca: VWR Collection
VWR® Socket Caps and Colour Inserts for Screw Microtubes
numer katalogowy 525-1671
Dostawca: VWR Collection
New Product
Przejdź do:
Product Details & Documents
Customisable socket caps with O-ring, accept colourful cap inserts to organise samples or match kit branding. For use with VWR® screw cap microtubes only.
Feature ¾ turn to easily open and close tubes requiring less revolutions than competitor tubes.
Dane techniczne
- Description:Customisable screw caps, non sterile
- Seq:4