Order Entry
Poland
Orders LinkContactUsLinkComponent
VWR® Socket Caps and Colour Inserts for Screw Microtubes | Avantor
VWR® Socket Caps and Colour Inserts for Screw Microtubes
Customisable screw caps, non sterile
numer katalogowy 525-1649
New Product
Dostawca:   VWR Collection
VWR® Socket Caps and Colour Inserts for Screw Microtubes
numer katalogowy 525-1649
Dostawca:  VWR Collection
New Product
VWR® Socket Caps and Colour Inserts for Screw Microtubes
VWR® Socket Caps and Colour Inserts for Screw Microtubes
Customisable screw caps, non sterile
Przejdź do:
Price
255 zł
Zaloguj się dla dostępności
1 * 500 SZT
Product Details & Documents
Customisable socket caps with O-ring, accept colourful cap inserts to organise samples or match kit branding. For use with VWR® screw cap microtubes only.
Feature ¾ turn to easily open and close tubes requiring less revolutions than competitor tubes.
Dane techniczne
  • Description:
    Customisable screw caps, non sterile
  • Seq:
    2