1-Hexadecanol ≥98%
Bottle
Catalog # APOSBUP19881-10G
New Product
Supplier: Apollo Scientific
1-Hexadecanol ≥98%
Catalog # APOSBUP19881-10G
Supplier: Apollo Scientific
New Product
CAS Number: 36653-82-4
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
- Non-hazardous
Synonyms: Cetyl alcohol, Palmityl alcohol
- CAS Number:36653-82-4
- Formula:C₁₆H₃₄O
- MDL Number:MFCD00004760
- Molecular Weight:242,44 g/mol
- EINECS:253-149-0
Specifications
Specifications
- Pack type:Bottle



