Złóż zamówienie
Poland
Orders LinkContactUsLinkComponent
 

Copper (II) Carbonate (basic) 53 - 57% (Cu basis) 53 - 57% (Cu basis)

New Product
Supplier:   Apollo Scientific
Copper (II) Carbonate (basic) 53 - 57% (Cu basis) 53 - 57% (Cu basis)
Supplier:  Apollo Scientific
New Product
Certificates
Copper (II) Carbonate (basic) 53 - 57% (Cu basis) 53 - 57% (Cu basis)

4 Options Available Below

Product Details & Documents

Usage: Inorganic salt or catalyst.

Synonyms:  Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)
  • CAS Number:12069-69-1
  • Formula:CuH₂O₂·Cu·CO₃
  • MDL Number:MFCD00010976
  • Molecular Weight:221,11558 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 107 zł
    107 zł
    1 * 100 g
       107 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 283 zł
    283 zł
    1 * 500 g
       283 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 280 zł
    1 280 zł
    1 * 2,5 kg
       1 280 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - 12 380 zł
    12 380 zł
    1 * 25 kg
       12 380 zł

    Specifications:
    New Product
    New Product