Order Entry
Poland
Orders LinkContactUsLinkComponent
Urea ≥95%

Urea ≥95%

New Product
Supplier:   Apollo Scientific
Urea ≥95%
Supplier:  Apollo Scientific
New Product
Certificates

2 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:CH₄N₂O
  • Molecular Weight:60,06 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 191 zł
    191 zł
    1 * 50 mg
       191 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 262 zł
    262 zł
    1 * 100 mg
       262 zł

    Specifications:
    New Product
    New Product