Order Entry
Poland
Orders LinkContactUsLinkComponent
Pterostilbene ≥98%

Pterostilbene ≥98%

Bottle
Catalog # APOSBUP00051-20MG
New Product
Supplier:   Apollo Scientific
Pterostilbene ≥98%
Catalog # APOSBUP00051-20MG
Supplier:  Apollo Scientific
New Product
CAS Number:  537-42-8
Pterostilbene ≥98%

Bottle

CAS Number:  537-42-8
Price
255 zł
Log in for availability
1 * 20 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  (E)-4-(3,5-Dimethoxystyryl)phenol, 4-[(E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol
  • CAS Number:537-42-8
  • Formula:C₁₆H₁₆O₃
  • Molecular Weight:256,3 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.