Order Entry
Poland
Orders LinkContactUsLinkComponent
Capsanthin antioxidant

Capsanthin antioxidant

Catalog # ABCAAB142638-50
Supplier:   Abcam
Capsanthin antioxidant
Catalog # ABCAAB142638-50
Supplier:  Abcam
CAS Number:  
Capsanthin antioxidant
CAS Number:  
Price
938 zł
Log in for availability
1 * 50 g
Product Details & Documents

Potent antioxidant. Main carotenoid in paprika. Active in vitro. Achieve your results faster with highly validated, pure and trusted compounds.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Paprika Extract
  • Molecular Weight:584,9 g/mol
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.