Order Entry
Poland
Orders LinkContactUsLinkComponent
FFN511 ≥98% Fluorescent substrate for VMAT2

FFN511 ≥98% Fluorescent substrate for VMAT2

Dostawca:   Abcam
FFN511 ≥98% Fluorescent substrate for VMAT2
Dostawca:  Abcam
Certyfikaty
FFN511 ≥98% Fluorescent substrate for VMAT2

2 dostępnych opcji poniżej

Product Details & Documents

First novel fluorescent false neurotransmitter. Enables optical imaging of presynaptic terminal activity. Inhibits 5-HT binding to VMAT2 (IC₅₀ = 1 μM (comparable to dopamine itself)). Released via exocytosis in the striatum in mouse acute slice preparation. Labels dopamine and other presynaptic terminals in the striatum. Compatible with GFP tags and other optical probes. Sufficiently bright, photostable and suitable for two-photon fluorescence microscopy. View the technique online For highly selective labelling of dopaminergic presynaptic termainals FFN102 is recommended.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonimy:  FFN-511
  • Numer CAS:1004548-96-2
  • Molecular Weight:284,35 g/mol
Specifications for Product Family

Wyniki testu specyfikacji

  • Constituents::
    100% FFN511
Product Family Options

Informacje o produkcie

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.